siRNA Details
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004501.3 | NP_004492.2 | 14 | 3602 | ||
NM_031844.2 | NP_114032.2 | 14 | 3659 | |||
XM_017001115.1 | XP_016856604.1 | 14 | 3854 | |||
XM_017001116.1 | XP_016856605.1 | 14 | 3659 | |||
XM_017001117.1 | XP_016856606.1 | 14 | 3648 | |||
GenBank mRNA | AK095525.1 | - | 1 | 185 | ||
AK126868.1 | - | 15 | 3400 | |||
BC015782.1 | - | 14 | 3136 | 4857112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
