siRNA Details
Gene Symbol: PLEKHG4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052909.3 | NP_443141.3 | 8, 9 | 1331 | ||
XM_006714446.3 | XP_006714509.1 | 10, 11 | 2469 | |||
XM_011513968.2 | XP_011512270.1 | 10, 11 | 2469 | |||
XM_011513969.2 | XP_011512271.1 | 10, 11 | 2346 | |||
XM_011513970.1 | XP_011512272.1 | 10, 11 | 2058 | |||
XM_011513971.2 | XP_011512273.1 | 10, 11 | 2414 | |||
XM_011513972.2 | XP_011512274.1 | 10, 11 | 2470 | |||
XM_011513973.1 | XP_011512275.1 | 10, 11 | 2467 | |||
XM_017009112.1 | XP_016864601.1 | 10, 11 | 2470 | |||
GenBank mRNA | AB067496.2 | - | 8, 9 | 1331 | ||
BC008352.1 | - | 8, 9 | 1023 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
