siRNA Details
Gene Symbol: PLEKHG4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052909.3 | NP_443141.3 | 9 | 1359 | ||
XM_006714446.3 | XP_006714509.1 | 11 | 2497 | |||
XM_011513968.2 | XP_011512270.1 | 11 | 2497 | |||
XM_011513969.2 | XP_011512271.1 | 11 | 2374 | |||
XM_011513970.1 | XP_011512272.1 | 11 | 2086 | |||
XM_011513971.2 | XP_011512273.1 | 11 | 2442 | |||
XM_011513972.2 | XP_011512274.1 | 11 | 2498 | |||
XM_011513973.1 | XP_011512275.1 | 11 | 2495 | |||
XM_017009112.1 | XP_016864601.1 | 11 | 2498 | |||
GenBank mRNA | AB067496.2 | - | 9 | 1359 | ||
BC008352.1 | - | 9 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
