siRNA Details
Gene Symbol: CMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030629.2 | NP_085132.1 | 3 | 302 | ||
NM_198390.2 | NP_938204.2 | 3 | 528 | |||
XM_005256179.4 | XP_005256236.4 | 3 | 1483 | |||
XM_005256181.2 | XP_005256238.1 | 2 | 259 | |||
XM_011523352.1 | XP_011521654.1 | 3 | 488 | |||
GenBank mRNA | AB051481.1 | - | 3 | 409 | ||
AK096598.1 | - | 3 | 489 | |||
AK304418.1 | - | 2 | 264 | |||
AK308869.1 | - | 2 | 264 | |||
AY172689.1 | - | 3 | 302 | |||
CV570964.1 | - | 3 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
