siRNA Details
Gene Symbol: CMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030629.2 | NP_085132.1 | 2 | 179 | ||
NM_198390.2 | NP_938204.2 | 2 | 405 | |||
XM_005256179.4 | XP_005256236.4 | 2 | 1360 | |||
XM_011523352.1 | XP_011521654.1 | 2 | 365 | |||
GenBank mRNA | AB051481.1 | - | 2 | 286 | ||
AK096598.1 | - | 2 | 366 | |||
AY172689.1 | - | 2 | 179 | |||
CV570964.1 | - | 2 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
