siRNA Details
Gene Symbol: CMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030629.2 | NP_085132.1 | 11 | 1278 | ||
NM_198390.2 | NP_938204.2 | 11 | 1504 | |||
XM_005256179.4 | XP_005256236.4 | 11 | 2459 | |||
XM_005256181.2 | XP_005256238.1 | 10 | 1235 | |||
XM_005256182.1 | XP_005256239.1 | 9 | 992 | |||
XM_011523352.1 | XP_011521654.1 | 11 | 1464 | |||
XM_011523353.1 | XP_011521655.1 | 9 | 1372 | |||
XM_017023733.1 | XP_016879222.1 | 9 | 5849 | |||
GenBank mRNA | AB051481.1 | - | 11 | 1385 | ||
AK096598.1 | - | 11 | 1465 | |||
AK304418.1 | - | 10 | 1240 | |||
AY172689.1 | - | 11 | 1278 | |||
BC038113.1 | - | 9 | 1201 | 4818904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
