siRNA Details
Gene Symbol: MCOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153259.3 | NP_694991.2 | 11 | 1568 | ||
XM_005270719.3 | XP_005270776.1 | 11 | 1345 | |||
XM_006710552.2 | XP_006710615.1 | 12 | 1658 | |||
XM_011541187.2 | XP_011539489.1 | 11 | 1402 | |||
XM_011541188.2 | XP_011539490.1 | 11 | 1685 | |||
GenBank mRNA | AK094010.1 | - | 9 | 1243 | ||
AK129786.1 | - | 5 | 571 | |||
AK309858.1 | - | 11 | 1559 | |||
AY083533.1 | - | 10 | 1234 | |||
BC104891.1 | - | 11 | 1386 | 8143894 | ||
BC104893.1 | - | 11 | 1386 | 8143896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
