siRNA Details
Gene Symbol: MCOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153259.3 | NP_694991.2 | 6 | 966 | ||
XM_005270719.3 | XP_005270776.1 | 6 | 743 | |||
XM_006710552.2 | XP_006710615.1 | 7 | 1056 | |||
XM_011541187.2 | XP_011539489.1 | 6 | 800 | |||
XM_011541188.2 | XP_011539490.1 | 6 | 1083 | |||
XM_017000923.1 | XP_016856412.1 | 6 | 957 | |||
GenBank mRNA | AK094010.1 | - | 6 | 957 | ||
AK309858.1 | - | 6 | 957 | |||
AY083533.1 | - | 5 | 632 | |||
BC104891.1 | - | 6 | 784 | 8143894 | ||
BC104893.1 | - | 6 | 784 | 8143896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
