siRNA Details
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138459.3 | NP_612468.1 | 5 | 2777 | ||
GenBank mRNA | AJ420576.1 | - | 1 | 1046 | ||
AK025467.1 | - | 1 | 1008 | |||
AK125119.1 | - | 5 | 2739 | |||
AK126335.1 | - | 6 | 2490 | |||
BC110325.1 | - | 5 | 2732 | 5553827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
