siRNA Details
Gene Symbol: NUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138459.3 | NP_612468.1 | 5 | 3244 | ||
GenBank mRNA | AJ420576.1 | - | 1 | 1513 | ||
AK025467.1 | - | 1 | 1475 | |||
AK125119.1 | - | 5 | 3206 | |||
AK126335.1 | - | 6 | 2957 | |||
BC110325.1 | - | 5 | 3199 | 5553827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
