siRNA Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 3 | 772 | ||
NM_018212.4 | NP_060682.2 | 3 | 772 | |||
XM_005273187.2 | XP_005273244.1 | 4 | 812 | |||
XM_011544229.2 | XP_011542531.2 | 3 | 1231 | |||
XM_017001746.1 | XP_016857235.1 | 3 | 1978 | |||
XM_017001747.1 | XP_016857236.1 | 3 | 1231 | |||
XM_017001748.1 | XP_016857237.1 | 3 | 1231 | |||
XM_017001749.1 | XP_016857238.1 | 3 | 1231 | |||
XM_017001750.1 | XP_016857239.1 | 3 | 1231 | |||
XM_017001751.1 | XP_016857240.1 | 3 | 1120 | |||
XM_017001752.1 | XP_016857241.1 | 3 | 1231 | |||
XM_017001753.1 | XP_016857242.1 | 4 | 782 | |||
XM_017001754.1 | XP_016857243.1 | 3 | 726 | |||
XM_017001755.1 | XP_016857244.1 | 3 | 469 | |||
XM_017001756.1 | XP_016857245.1 | 3 | 469 | |||
XM_017001757.1 | XP_016857246.1 | 3 | 372 | |||
GenBank mRNA | AF519769.1 | - | 3 | 588 | ||
AK096246.1 | - | 1 | 68 | |||
AK290179.1 | - | 3 | 716 | |||
AY345143.1 | - | 3 | 676 | |||
BC065238.1 | - | 2 | 190 | |||
BC095481.1 | - | 3 | 771 | 30348815 | ||
EU255274.1 | - | 2 | 323 | |||
KM214203.1 | - | 2 | 319 | |||
KM214204.1 | - | 2 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
