siRNA Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 15 | 3148 | ||
NM_018212.4 | NP_060682.2 | 14 | 3085 | |||
XM_005273187.2 | XP_005273244.1 | 16 | 3188 | |||
XM_011544229.2 | XP_011542531.2 | 16 | 3664 | |||
XM_017001746.1 | XP_016857235.1 | 17 | 4423 | |||
XM_017001747.1 | XP_016857236.1 | 16 | 3613 | |||
XM_017001748.1 | XP_016857237.1 | 15 | 3607 | |||
XM_017001749.1 | XP_016857238.1 | 15 | 3601 | |||
XM_017001750.1 | XP_016857239.1 | 16 | 3565 | |||
XM_017001751.1 | XP_016857240.1 | 14 | 3433 | |||
XM_017001752.1 | XP_016857241.1 | 13 | 3433 | |||
XM_017001753.1 | XP_016857242.1 | 18 | 3227 | |||
XM_017001754.1 | XP_016857243.1 | 17 | 3171 | |||
XM_017001755.1 | XP_016857244.1 | 17 | 2914 | |||
XM_017001756.1 | XP_016857245.1 | 15 | 2845 | |||
XM_017001757.1 | XP_016857246.1 | 15 | 2748 | |||
GenBank mRNA | AF519769.1 | - | 15 | 2962 | ||
AK001635.1 | - | 8 | 1565 | |||
AK096246.1 | - | 12 | 2381 | |||
AK126894.1 | - | 9 | 1842 | |||
AK127561.1 | - | 9 | 2278 | |||
AL133059.1 | - | 8 | 1456 | |||
BC095481.1 | - | 15 | 3147 | 30348815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
