siRNA Details
Gene Symbol: FLCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144997.5 | NP_659434.2 | 10, 11 | 1679 | ||
XM_011523714.2 | XP_011522016.1 | 11, 12 | 1730 | |||
XM_011523716.1 | XP_011522018.1 | 9, 10 | 2108 | |||
XM_011523718.2 | XP_011522020.1 | 10, 11 | 1614 | |||
XM_011523719.2 | XP_011522021.1 | 11, 12 | 1730 | |||
XM_011523721.2 | XP_011522023.1 | 10, 11 | 1479 | |||
XM_017024304.1 | XP_016879793.1 | 12, 13 | 1899 | |||
XM_017024305.1 | XP_016879794.1 | 12, 13 | 1914 | |||
XM_017024306.1 | XP_016879795.1 | 11, 12 | 1903 | |||
XM_017024307.1 | XP_016879796.1 | 11, 12 | 1865 | |||
XM_017024308.1 | XP_016879797.1 | 12, 13 | 2041 | |||
XM_017024309.1 | XP_016879798.1 | 9, 10 | 1454 | |||
GenBank mRNA | AF517523.1 | - | 10, 11 | 1630 | ||
AK309336.1 | - | 6, 7 | 798 | |||
BC015725.2 | - | 3, 4 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
