siRNA Details
Gene Symbol: FLCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144997.5 | NP_659434.2 | 14 | 2559 | ||
XM_011523714.2 | XP_011522016.1 | 15 | 2610 | |||
XM_011523716.1 | XP_011522018.1 | 13 | 2988 | |||
XM_011523718.2 | XP_011522020.1 | 14 | 2494 | |||
XM_011523721.2 | XP_011522023.1 | 14 | 2359 | |||
XM_017024304.1 | XP_016879793.1 | 16 | 2779 | |||
XM_017024305.1 | XP_016879794.1 | 16 | 2794 | |||
XM_017024306.1 | XP_016879795.1 | 15 | 2783 | |||
XM_017024307.1 | XP_016879796.1 | 15 | 2745 | |||
XM_017024309.1 | XP_016879798.1 | 13 | 2334 | |||
GenBank mRNA | AF517523.1 | - | 14 | 2510 | ||
BC015725.2 | - | 7 | 1259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
