siRNA Details
Gene Symbol: CNTNAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130773.3 | NP_570129.1 | 8 | 1642 | ||
XM_006712258.1 | XP_006712321.1 | 8 | 1645 | |||
XM_017003316.1 | XP_016858805.1 | 8 | 1645 | |||
GenBank mRNA | AB077881.1 | - | 8 | 1278 | ||
AK056528.1 | - | 8 | 1645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
