siRNA Details
Gene Symbol: CCDC146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020879.2 | NP_065930.2 | 13 | 1670 | ||
GenBank mRNA | AB040938.1 | - | 6 | 2772 | ||
AK056542.1 | - | 12 | 1448 | |||
AK292491.1 | - | 13 | 1649 | |||
BC036053.1 | - | 13 | 1670 | 5273288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
