siRNA Details
Gene Symbol: CCDC146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020879.2 | NP_065930.2 | 19 | 2936 | ||
GenBank mRNA | AB040938.1 | - | 12 | 4038 | ||
AF461817.1 | - | 5 | 1147 | |||
AK056542.1 | - | 18 | 2714 | |||
AK292491.1 | - | 19 | 2915 | |||
BC029458.1 | - | 5 | 964 | |||
BC036053.1 | - | 19 | 2936 | 5273288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
