siRNA Details
Gene Symbol: CCDC146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020879.2 | NP_065930.2 | 16 | 2348 | ||
GenBank mRNA | AB040938.1 | - | 9 | 3450 | ||
AF461817.1 | - | 2 | 559 | |||
AK056542.1 | - | 15 | 2126 | |||
AK292491.1 | - | 16 | 2327 | |||
BC029458.1 | - | 2 | 376 | |||
BC036053.1 | - | 16 | 2348 | 5273288 | ||
HM370402.1 | - | 5 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
