siRNA Details
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 7 | 2657 | ||
NM_001270527.1 | NP_001257456.1 | 6 | 807 | |||
NM_004099.5 | NP_004090.4 | 7 | 2748 | |||
NM_198194.2 | NP_937837.1 | 3 | 2253 | |||
NR_073037.1 | 8 | 2878 | ||||
GenBank mRNA | AL040491.1 | - | 1 | 55 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
