siRNA Details
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001186.3 | NP_001177.1 | 3 | 848 | ||
NM_206866.2 | NP_996749.1 | 3 | 872 | |||
NR_027655.2 | 3 | 808 | ||||
GenBank mRNA | AB002803.1 | - | 3 | 747 | ||
AF026199.1 | - | 3 | 872 | |||
AK291021.1 | - | 3 | 805 | |||
AK313791.1 | - | 3 | 721 | |||
BC063307.1 | - | 3 | 721 | 30340811 | ||
CR450303.1 | - | 2 | 629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
