siRNA Details
Gene Symbol: PCDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278613.1 | NP_001265542.1 | 2 | 385 | ||
NM_002587.4 | NP_002578.2 | 2 | 462 | |||
NM_032420.3 | NP_115796.2 | 2 | 462 | |||
XM_005268452.2 | XP_005268509.1 | 2 | 2792 | |||
XM_005268454.4 | XP_005268511.1 | 2 | 2792 | |||
GenBank mRNA | AK289512.1 | - | 2 | 462 | ||
AK304231.1 | - | 2 | 347 | |||
BC035812.1 | - | 2 | 431 | 5753628 | ||
DA096772.1 | - | 2 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
