siRNA Details
Gene Symbol: PCDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278613.1 | NP_001265542.1 | 3 | 2107 | ||
NM_001278615.1 | NP_001265544.1 | 2 | 1264 | |||
NM_002587.4 | NP_002578.2 | 3 | 2184 | |||
NM_032420.3 | NP_115796.2 | 3 | 2184 | |||
XM_005268452.2 | XP_005268509.1 | 3 | 4514 | |||
XM_005268454.4 | XP_005268511.1 | 3 | 4514 | |||
XM_005268455.2 | XP_005268512.1 | 2 | 1207 | |||
XM_017009517.1 | XP_016865006.1 | 2 | 2141 | |||
XM_017009518.1 | XP_016865007.1 | 2 | 1207 | |||
GenBank mRNA | AK075233.1 | - | 2 | 1553 | ||
AK289512.1 | - | 3 | 2184 | |||
AK300564.1 | - | 2 | 1207 | |||
AK304231.1 | - | 3 | 2069 | |||
AK308338.1 | - | 2 | 1264 | |||
BC035812.1 | - | 3 | 2153 | 5753628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
