siRNA Details
Gene Symbol: FBXO32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058229.3 | NP_478136.1 | 6 | 828 | ||
NM_148177.2 | NP_680482.1 | 3 | 463 | |||
GenBank mRNA | AJ420108.1 | - | 6 | 678 | ||
AK056986.1 | - | 6 | 781 | |||
AY059629.1 | - | 6 | 589 | |||
BC024030.2 | - | 3 | 463 | 4825638 | ||
BC100020.1 | - | 5 | 419 | |||
BC120963.1 | - | 3 | 457 | 40115317 | ||
BC120964.1 | - | 3 | 457 | 40115318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
