siRNA Details
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 31 | 4083 | ||
GenBank mRNA | X57398.1 | - | 31 | 3921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
