siRNA Details
Gene Symbol: ATG16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190266.1 | NP_001177195.1 | 12 | 1398 | ||
NM_001190267.1 | NP_001177196.1 | 12 | 1396 | |||
NM_017974.3 | NP_060444.3 | 11 | 1343 | |||
NM_030803.6 | NP_110430.5 | 12 | 1400 | |||
NM_198890.2 | NP_942593.2 | 8 | 911 | |||
XM_005246082.2 | XP_005246139.1 | 13 | 1388 | |||
XM_005246084.2 | XP_005246141.1 | 10 | 927 | |||
XM_005246086.2 | XP_005246143.1 | 9 | 876 | |||
XM_006712608.2 | XP_006712671.1 | 12 | 1199 | |||
GenBank mRNA | AK000897.1 | - | 11 | 1011 | ||
AK001690.1 | - | 11 | 1188 | |||
AK024453.1 | - | 11 | 1938 | |||
AK027854.1 | - | 10 | 925 | |||
AK123876.1 | - | 9 | 762 | |||
AK308400.1 | - | 11 | 1241 | |||
AY358182.1 | - | 12 | 1289 | |||
AY398617.1 | - | 12 | 1289 | |||
BC071846.1 | - | 8 | 749 | |||
BC117337.1 | - | 12 | 1062 | |||
BC143900.1 | - | 11 | 1005 | |||
EF079889.1 | - | 11 | 1086 | |||
EF079890.1 | - | 8 | 654 | |||
EF560734.1 | - | 11 | 1213 | |||
JQ924062.1 | - | 11 | 1086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
