siRNA Details
Gene Symbol: ATG16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190266.1 | NP_001177195.1 | 12 | 1444 | ||
NM_001190267.1 | NP_001177196.1 | 12 | 1442 | |||
NM_017974.3 | NP_060444.3 | 11 | 1389 | |||
NM_030803.6 | NP_110430.5 | 12 | 1446 | |||
NM_198890.2 | NP_942593.2 | 8 | 957 | |||
XM_005246082.2 | XP_005246139.1 | 13 | 1434 | |||
XM_005246084.2 | XP_005246141.1 | 10 | 973 | |||
XM_005246086.2 | XP_005246143.1 | 9 | 922 | |||
XM_006712608.2 | XP_006712671.1 | 12 | 1245 | |||
GenBank mRNA | AK000897.1 | - | 11 | 1057 | ||
AK001690.1 | - | 11 | 1234 | |||
AK024453.1 | - | 11 | 1984 | |||
AK027854.1 | - | 10 | 971 | |||
AK074434.1 | - | 6 | 416 | |||
AK123876.1 | - | 9 | 808 | |||
AK308400.1 | - | 11 | 1287 | |||
AY358182.1 | - | 12 | 1335 | |||
AY398617.1 | - | 12 | 1335 | |||
BC071846.1 | - | 8 | 795 | |||
BC117337.1 | - | 12 | 1108 | |||
BC143900.1 | - | 11 | 1051 | |||
EF079889.1 | - | 11 | 1132 | |||
EF079890.1 | - | 8 | 700 | |||
EF560734.1 | - | 11 | 1259 | |||
JQ924062.1 | - | 11 | 1132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
