siRNA Details
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 3 | 1102 | ||
NM_001256062.2 | NP_001242991.1 | 2 | 1236 | |||
NM_001256321.2 | NP_001243250.1 | 3 | 837 | |||
NM_001256322.2 | NP_001243251.1 | 2 | 1231 | |||
NM_001256323.2 | NP_001243252.1 | 3 | 832 | |||
NM_024328.5 | NP_077304.1 | 2 | 1501 | |||
NR_023314.4 | 2 | 540 | ||||
NR_046051.1 | 5 | 643 | ||||
NR_046052.1 | 3 | 423 | ||||
GenBank mRNA | AF432862.1 | - | 2 | 770 | ||
AK057691.1 | - | 2 | 1208 | |||
AK098604.1 | - | 3 | 891 | |||
BC002984.1 | - | 2 | 1195 | 3535451 | ||
BX161435.1 | - | 2 | 1461 | |||
CB054399.1 | - | 2 | 479 | |||
DA952722.1 | - | 3 | 546 | |||
DB044416.1 | - | 2 | 537 | |||
DB257923.1 | - | 2 | 536 | |||
DT220066.1 | - | 2 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
