siRNA Details
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 2 | 902 | ||
NM_024328.5 | NP_077304.1 | 1 | 1301 | |||
GenBank mRNA | AF432862.1 | - | 1 | 570 | ||
AK057691.1 | - | 1 | 1008 | |||
AK098604.1 | - | 2 | 691 | |||
BC002984.1 | - | 1 | 995 | 3535451 | ||
BI522644.1 | - | 2 | 706 | |||
BX161435.1 | - | 1 | 1261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
