siRNA Details
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 3 | 1440 | ||
NM_001256062.2 | NP_001242991.1 | 2 | 1574 | |||
NM_001256321.2 | NP_001243250.1 | 3 | 1175 | |||
NM_001256322.2 | NP_001243251.1 | 2 | 1569 | |||
NM_001256323.2 | NP_001243252.1 | 3 | 1170 | |||
NM_024328.5 | NP_077304.1 | 2 | 1839 | |||
NR_023314.4 | 2 | 878 | ||||
NR_046051.1 | 5 | 981 | ||||
NR_046052.1 | 3 | 761 | ||||
GenBank mRNA | AK057691.1 | - | 2 | 1546 | ||
AK094211.1 | - | 3 | 754 | |||
AK098604.1 | - | 3 | 1229 | |||
BC002984.1 | - | 2 | 1533 | 3535451 | ||
BU535676.1 | - | 1 | 329 | |||
BX161435.1 | - | 2 | 1799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
