siRNA Details
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004965.6 | NP_004956.5 | 6 | 529 | ||
XM_011529543.1 | XP_011527845.1 | 9 | 1256 | |||
XM_011529544.1 | XP_011527846.1 | 8 | 1210 | |||
XM_011529545.1 | XP_011527847.1 | 8 | 590 | |||
XM_011529547.1 | XP_011527849.1 | 6 | 439 | |||
XM_011529549.1 | XP_011527851.1 | 8 | 751 | |||
XM_017028332.1 | XP_016883821.1 | 8 | 2040 | |||
XM_017028333.1 | XP_016883822.1 | 9 | 2112 | |||
XM_017028334.1 | XP_016883823.1 | 8 | 2022 | |||
XM_017028335.1 | XP_016883824.1 | 7 | 544 | |||
XM_017028336.1 | XP_016883825.1 | 8 | 673 | |||
GenBank mRNA | AB209245.1 | - | 5 | 3482 | ||
AK056033.1 | - | 3 | 899 | |||
AK130775.1 | - | 2 | 110 | |||
BC000075.2 | - | 6 | 449 | 3510312 | ||
BC023984.1 | - | 6 | 441 | 4104558 | ||
BC070153.1 | - | 6 | 467 | 6450636 | ||
BC070154.1 | - | 6 | 447 | 6495327 | ||
BC071643.1 | - | 6 | 437 | 5549431 | ||
BC106080.1 | - | 6 | 462 | 3855939 | ||
BC107741.1 | - | 6 | 535 | 6495117 | ||
BE779939.1 | - | 589 | ||||
J02621.1 | - | 6 | 475 |
Gene Symbol: ACTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641076.1 | - | 1 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
