siRNA Details
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004965.6 | NP_004956.5 | 6 | 1057 | ||
XM_011529543.1 | XP_011527845.1 | 9 | 1784 | |||
XM_011529544.1 | XP_011527846.1 | 8 | 1738 | |||
XM_011529545.1 | XP_011527847.1 | 8 | 1118 | |||
XM_011529547.1 | XP_011527849.1 | 6 | 967 | |||
XM_011529549.1 | XP_011527851.1 | 8 | 1279 | |||
XM_017028332.1 | XP_016883821.1 | 8 | 2568 | |||
XM_017028333.1 | XP_016883822.1 | 9 | 2640 | |||
XM_017028334.1 | XP_016883823.1 | 8 | 2550 | |||
XM_017028335.1 | XP_016883824.1 | 7 | 1072 | |||
XM_017028336.1 | XP_016883825.1 | 8 | 1201 | |||
GenBank mRNA | AK130775.1 | - | 2 | 638 | ||
BC106080.1 | - | 6 | 990 | 3855939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
