siRNA Details
Gene Symbol: CABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033677.1 | NP_001028849.1 | 4 | 966 | ||
NM_004276.4 | NP_004267.2 | 4 | 421 | |||
NM_031205.3 | NP_112482.1 | 5 | 601 | |||
XM_017020235.1 | XP_016875724.1 | 4 | 1033 | |||
XM_017020236.1 | XP_016875725.1 | 4 | 1168 | |||
XM_017020237.1 | XP_016875726.1 | 4 | 1168 | |||
XM_017020238.1 | XP_016875727.1 | 5 | 2247 | |||
XM_017020239.1 | XP_016875728.1 | 4 | 2061 | |||
GenBank mRNA | AF169148.1 | - | 4 | 360 | ||
AF169149.1 | - | 5 | 540 | |||
AK127203.1 | - | 2 | 1435 | |||
AL533429.3 | - | 4 | 936 | |||
BC030201.1 | - | 4 | 966 | 5200625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
