siRNA Details
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 26 | 4528 | ||
XM_005246263.2 | XP_005246320.1 | 27 | 4689 | |||
XM_005246265.3 | XP_005246322.1 | 27 | 4642 | |||
XM_011510530.1 | XP_011508832.1 | 27 | 4616 | |||
XM_011510531.1 | XP_011508833.1 | 28 | 4710 | |||
XM_011510532.2 | XP_011508834.1 | 28 | 4742 | |||
XM_011510533.2 | XP_011508835.1 | 28 | 4663 | |||
XM_011510534.1 | XP_011508836.1 | 26 | 4547 | |||
XM_011510535.2 | XP_011508837.1 | 26 | 4463 | |||
XM_011510536.2 | XP_011508838.1 | 21 | 3567 | |||
GenBank mRNA | AB067489.1 | - | 26 | 4299 | ||
AK094865.1 | - | 10 | 2083 | |||
AK131316.1 | - | 13 | 2820 | |||
AL834396.1 | - | 24 | 3912 | |||
BC014343.1 | - | 1 | 605 | |||
BC167159.1 | - | 26 | 4299 | 8862581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
