siRNA Details
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 22 | 3119 | ||
XM_005246263.2 | XP_005246320.1 | 22 | 3186 | |||
XM_005246265.3 | XP_005246322.1 | 22 | 3186 | |||
XM_011510530.1 | XP_011508832.1 | 23 | 3207 | |||
XM_011510531.1 | XP_011508833.1 | 23 | 3207 | |||
XM_011510532.2 | XP_011508834.1 | 23 | 3207 | |||
XM_011510533.2 | XP_011508835.1 | 23 | 3207 | |||
XM_011510534.1 | XP_011508836.1 | 22 | 3138 | |||
XM_011510535.2 | XP_011508837.1 | 22 | 3054 | |||
XM_011510536.2 | XP_011508838.1 | 17 | 2158 | |||
GenBank mRNA | AB067489.1 | - | 22 | 2890 | ||
AK094865.1 | - | 6 | 674 | |||
AK131316.1 | - | 8 | 1317 | |||
AL834396.1 | - | 20 | 2503 | |||
BC113878.1 | - | 9 | 1081 | |||
BC114438.1 | - | 9 | 1086 | |||
BC167159.1 | - | 22 | 2890 | 8862581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
