siRNA Details
Gene Symbol: TMSB15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021992.2 | NP_068832.1 | 3 | 543 | ||
GenBank mRNA | AF086495.1 | - | 3 | 486 | ||
BC000183.1 | - | 3 | 491 | 3350388 | ||
BG829126.1 | - | 3 | 493 | |||
BU599402.1 | - | 3 | 556 | |||
D82345.1 | - | 3 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
