siRNA Details
Gene Symbol: PCDHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018909.3 | NP_061732.1 | 4 | 3534 | ||
NM_031849.2 | NP_114037.1 | 4 | 2742 | |||
GenBank mRNA | AB209205.1 | - | 4 | 2310 | ||
AF152314.1 | - | 4 | 3364 | |||
BC036674.2 | - | 4 | 3478 | 5263226 |
Gene Symbol: PCDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018900.3 | NP_061723.1 | 4 | 3519 | ||
NM_031411.2 | NP_113599.1 | 4 | 2727 | |||
GenBank mRNA | AF152305.1 | - | 4 | 3364 |
Gene Symbol: PCDHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018901.3 | NP_061724.1 | 4 | 3524 | ||
NM_031860.2 | NP_114066.1 | 4 | 2735 | |||
GenBank mRNA | AF152306.1 | - | 4 | 3358 | ||
BC130447.1 | - | 4 | 2608 | 40146478 |
Gene Symbol: PCDHA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018902.4 | NP_061725.1 | 4 | 4952 | ||
GenBank mRNA | AF152307.1 | - | 4 | 3361 |
Gene Symbol: PCDHA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018903.3 | NP_061726.1 | 4 | 3508 | ||
GenBank mRNA | AF152308.1 | - | 4 | 3337 | ||
AK075556.1 | - | 4 | 2389 |
Gene Symbol: PCDHA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018904.2 | NP_061727.1 | 4 | 3364 | ||
GenBank mRNA | AB208924.1 | - | 5 | 2988 | ||
AF152309.1 | - | 4 | 3364 |
Gene Symbol: PCDHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018905.2 | NP_061728.1 | 4 | 3464 | ||
GenBank mRNA | AF152310.1 | - | 4 | 3358 |
Gene Symbol: PCDHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018906.2 | NP_061729.1 | 4 | 3364 | ||
GenBank mRNA | AF152311.1 | - | 4 | 3364 |
Gene Symbol: PCDHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018907.3 | NP_061730.1 | 4 | 3469 | ||
GenBank mRNA | AB208925.1 | - | 5 | 3032 | ||
AF152312.1 | - | 4 | 3355 |
Gene Symbol: PCDHA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018908.2 | NP_061731.1 | 4 | 3322 | ||
GenBank mRNA | AF152313.1 | - | 4 | 3322 | ||
BC033735.1 | - | 5 | 3024 | 5175471 |
Gene Symbol: PCDHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018910.2 | NP_061733.1 | 4 | 3325 | ||
GenBank mRNA | AF152315.1 | - | 4 | 3325 |
Gene Symbol: PCDHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018911.2 | NP_061734.1 | 4 | 3364 | ||
GenBank mRNA | AF152316.1 | - | 4 | 3364 |
Gene Symbol: PCDHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031857.1 | NP_114063.1 | 4 | 4088 | ||
GenBank mRNA | AF152317.1 | - | 4 | 3364 |
Gene Symbol: PCDHAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018898.3 | NP_061721.2 | 4 | 3579 | ||
GenBank mRNA | AF152303.1 | - | 4 | 3403 |
Gene Symbol: PCDHAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018899.5 | NP_061722.1 | 4 | 4140 | ||
GenBank mRNA | AF152304.1 | - | 4 | 3535 | ||
BC063288.1 | - | 4 | 3777 | 30343089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
