siRNA Details
Gene Symbol: PCDHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018911.2 | NP_061734.1 | 4 | 5182 | ||
GenBank mRNA | AF152316.1 | - | 4 | 5182 |
Gene Symbol: PCDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018900.3 | NP_061723.1 | 4 | 5337 | ||
NM_031411.2 | NP_113599.1 | 4 | 4545 | |||
GenBank mRNA | AF152305.1 | - | 4 | 5182 |
Gene Symbol: PCDHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018901.3 | NP_061724.1 | 4 | 5342 | ||
NM_031860.2 | NP_114066.1 | 4 | 4553 | |||
GenBank mRNA | AF152306.1 | - | 4 | 5176 |
Gene Symbol: PCDHA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018902.4 | NP_061725.1 | 4 | 6770 | ||
GenBank mRNA | AF152307.1 | - | 4 | 5179 |
Gene Symbol: PCDHA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018903.3 | NP_061726.1 | 4 | 5326 | ||
GenBank mRNA | AF152308.1 | - | 4 | 5155 |
Gene Symbol: PCDHA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018904.2 | NP_061727.1 | 4 | 5182 | ||
GenBank mRNA | AB208924.1 | - | 5 | 4806 | ||
AF152309.1 | - | 4 | 5182 |
Gene Symbol: PCDHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018905.2 | NP_061728.1 | 4 | 5282 | ||
GenBank mRNA | AF152310.1 | - | 4 | 5176 |
Gene Symbol: PCDHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018906.2 | NP_061729.1 | 4 | 5182 | ||
GenBank mRNA | AF152311.1 | - | 4 | 5182 |
Gene Symbol: PCDHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018907.3 | NP_061730.1 | 4 | 5287 | ||
GenBank mRNA | AF152312.1 | - | 4 | 5173 |
Gene Symbol: PCDHA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018908.2 | NP_061731.1 | 4 | 5140 | ||
GenBank mRNA | AF152313.1 | - | 4 | 5140 |
Gene Symbol: PCDHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018909.3 | NP_061732.1 | 4 | 5352 | ||
NM_031849.2 | NP_114037.1 | 4 | 4560 | |||
GenBank mRNA | AB209205.1 | - | 4 | 4128 | ||
AF152314.1 | - | 4 | 5182 |
Gene Symbol: PCDHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018910.2 | NP_061733.1 | 4 | 5143 | ||
GenBank mRNA | AF152315.1 | - | 4 | 5143 |
Gene Symbol: PCDHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031857.1 | NP_114063.1 | 4 | 5906 | ||
GenBank mRNA | AF152317.1 | - | 4 | 5182 |
Gene Symbol: PCDHAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018898.3 | NP_061721.2 | 4 | 5397 | ||
GenBank mRNA | AF152303.1 | - | 4 | 5221 |
Gene Symbol: PCDHAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018899.5 | NP_061722.1 | 4 | 5958 | ||
GenBank mRNA | AF152304.1 | - | 4 | 5353 | ||
BC063288.1 | - | 4 | 5595 | 30343089 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198117, NM_009960, NM_138663, NM_138661, NM_001003671, and others.
Gene Ontology Categories:
