siRNA Details
Gene Symbol: HIST1H2BD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021063.3 | NP_066407.1 | 1 | 423 | ||
NM_138720.2 | NP_619790.1 | 1 | 423 | |||
XM_005249039.3 | XP_005249096.1 | 1 | 559 | |||
GenBank mRNA | AJ223353.1 | - | 1 | 402 | ||
AK292712.1 | - | 1 | 419 | |||
BC002842.2 | - | 1 | 399 | 3659807 | ||
BC096122.3 | - | 1 | 374 | 40002354 | ||
BM153714.1 | - | 1 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
