siRNA Details
Gene Symbol: ACSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005622.3 | NP_005613.2 | 6 | 995 | ||
NM_202000.2 | NP_973729.1 | 6 | 995 | |||
XM_011545911.1 | XP_011544213.1 | 7 | 1100 | |||
XM_017023523.1 | XP_016879012.1 | 6 | 989 | |||
XM_017023524.1 | XP_016879013.1 | 5 | 859 | |||
GenBank mRNA | AK223139.1 | - | 6 | 964 | ||
AK290322.1 | - | 6 | 964 | |||
AK296122.1 | - | 6 | 794 | |||
BC002790.2 | - | 5 | 854 | 3617826 | ||
D16350.1 | - | 5 | 835 | |||
X80062.1 | - | 6 | 944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
