siRNA Details
Gene Symbol: ACSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005622.3 | NP_005613.2 | 3 | 556 | ||
NM_202000.2 | NP_973729.1 | 3 | 556 | |||
XM_011545911.1 | XP_011544213.1 | 3 | 550 | |||
XM_017023523.1 | XP_016879012.1 | 3 | 550 | |||
XM_017023524.1 | XP_016879013.1 | 2 | 420 | |||
GenBank mRNA | AK223139.1 | - | 3 | 525 | ||
AK290322.1 | - | 3 | 525 | |||
AK296122.1 | - | 2 | 244 | |||
BC002790.2 | - | 2 | 415 | 3617826 | ||
BI770558.1 | - | 3 | 556 | |||
D16350.1 | - | 2 | 396 | |||
X80062.1 | - | 3 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
