siRNA Details
Gene Symbol: USF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276373.1 | NP_001263302.1 | 3 | 136 | ||
NM_007122.4 | NP_009053.1 | 3 | 232 | |||
NM_207005.2 | NP_996888.1 | 3 | 232 | |||
GenBank mRNA | AB098540.1 | - | 1 | 25 | ||
AK314876.1 | - | 3 | 152 | |||
AL832119.1 | - | 3 | 269 | |||
BC035505.2 | - | 3 | 180 | 4559819 | ||
BP280296.1 | - | 3 | 230 | |||
KU178379.1 | - | 1 | 25 | |||
KU178380.1 | - | 1 | 25 | |||
X55666.1 | - | 3 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
