siRNA Details
Gene Symbol: WFDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130896.2 | NP_570966.2 | 4 | 386 | ||
NM_181510.2 | NP_852611.2 | 4 | 386 | |||
XM_017028119.1 | XP_016883608.1 | 4 | 365 | |||
GenBank mRNA | AF492015.1 | - | 4 | 386 | ||
AF492016.1 | - | 4 | 386 | |||
AL591715.1 | - | 3 | 281 | |||
GQ472209.1 | - | 4 | 386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
