siRNA Details
Gene Symbol: SLC25A37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317812.1 | NP_001304741.1 | 1, 2 | 417 | ||
NM_016612.3 | NP_057696.2 | 1, 2 | 417 | |||
GenBank mRNA | AF223466.1 | - | 1, 2 | 253 | ||
AK074708.1 | - | 1, 2 | 256 | |||
AK075313.1 | - | 1, 2 | 182 | |||
AK223194.1 | - | 1, 2 | 258 | |||
AL833186.1 | - | 1, 2 | 159 | |||
AY032628.1 | - | 1, 2 | 253 | |||
BC015013.2 | - | 1, 2 | 222 | |||
BC132799.1 | - | 1, 2 | 281 | 40146821 | ||
BC132801.1 | - | 1, 2 | 281 | 40146823 | ||
BI460337.1 | - | 1, 2 | 371 | |||
BM926580.1 | - | 1, 2 | 260 | |||
DB057758.1 | - | 1, 2 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
