siRNA Details
Gene Symbol: SLC25A37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317812.1 | NP_001304741.1 | 3, 4 | 1174 | ||
NM_001317813.1 | NP_001304742.1 | 4, 5 | 821 | |||
NM_001317814.1 | NP_001304743.1 | 4, 5 | 768 | |||
NM_016612.3 | NP_057696.2 | 3, 4 | 697 | |||
XM_006716352.3 | XP_006716415.1 | 3, 4 | 301 | |||
XM_011544550.2 | XP_011542852.1 | 3, 4 | 301 | |||
XM_011544553.2 | XP_011542855.1 | 4, 5 | 1533 | |||
XM_011544554.2 | XP_011542856.1 | 3, 4 | 2455 | |||
XM_017013532.1 | XP_016869021.1 | 3, 4 | 1372 | |||
GenBank mRNA | AF113696.1 | - | 1, 2 | 498 | ||
AF116630.1 | - | 1, 2 | 1320 | |||
AF155660.1 | - | 1, 2 | 515 | |||
AF223466.1 | - | 3, 4 | 533 | |||
AK074708.1 | - | 3, 4 | 1013 | |||
AK075313.1 | - | 3, 4 | 938 | |||
AK127666.1 | - | 3, 4 | 2766 | |||
AK223194.1 | - | 3, 4 | 1014 | |||
AY032628.1 | - | 2, 3 | 2520 | |||
BC132799.1 | - | 3, 4 | 561 | 40146821 | ||
BC132801.1 | - | 3, 4 | 561 | 40146823 | ||
BI460337.1 | - | 3, 4 | 651 | |||
BM552840.1 | - | 4, 5 | 703 | |||
BM926580.1 | - | 3, 4 | 540 | |||
BU166030.1 | - | 4, 5 | 741 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026331, AK050414, AK028588, AK162615, BC040399, and others.
Gene Ontology Categories:
