siRNA Details
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 7, 8 | 942 | ||
GenBank mRNA | AK092382.1 | - | 9, 10 | 737 | ||
AK315798.1 | - | 7, 8 | 811 | |||
BC005216.1 | - | 7, 8 | 816 | 3949903 | ||
BX497562.1 | - | 12 | ||||
M16630.1 | - | 7, 8 | 787 | |||
M18692.1 | - | 7, 8 | 814 |
Gene Symbol: CELA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005747.4 | NP_005738.4 | 7, 8 | 815 | ||
GenBank mRNA | AK225620.1 | - | 7, 8 | 811 | ||
BC005918.1 | - | 7, 8 | 815 | 4295129 | ||
BC007028.1 | - | 7, 8 | 816 | 3950453 | ||
BC008383.1 | - | 7, 8 | 811 | 3949901 | ||
BC015103.1 | - | 7, 8 | 819 | 3950076 | ||
BI833796.1 | - | 2, 3 | 265 | |||
D00306.1 | - | 7, 8 | 817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
