siRNA Details
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005159.4 | NP_005150.1 | 7 | 1414 | ||
GenBank mRNA | AK056592.1 | - | 7 | 954 | ||
AK129892.1 | - | 7 | 1237 | |||
AK129902.1 | - | 7 | 1237 | |||
AK290616.1 | - | 7 | 1237 | |||
BC009978.2 | - | 7 | 1376 | 4121697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
