siRNA Details
Gene Symbol: ARHGAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282290.1 | NP_001269219.1 | 1 | 491 | ||
NM_174919.3 | NP_777579.2 | 4 | 922 | |||
XM_006721745.2 | XP_006721808.1 | 4 | 873 | |||
XM_006721747.2 | XP_006721810.1 | 4 | 873 | |||
XM_006721750.3 | XP_006721813.1 | 4 | 846 | |||
XM_011524468.1 | XP_011522770.1 | 4 | 873 | |||
XM_011524469.1 | XP_011522771.1 | 4 | 873 | |||
XM_011524470.2 | XP_011522772.1 | 4 | 846 | |||
XM_011524471.1 | XP_011522773.1 | 4 | 873 | |||
XM_011524473.2 | XP_011522775.1 | 4 | 846 | |||
XM_017024311.1 | XP_016879800.1 | 2 | 807 | |||
XM_017024313.1 | XP_016879802.1 | 4 | 846 | |||
GenBank mRNA | AF258593.1 | - | 5 | 1864 | ||
AK290650.1 | - | 4 | 873 | |||
BC067345.1 | - | 4 | 922 | 6084901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
