siRNA Details
Gene Symbol: TRMU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282782.1 | NP_001269711.1 | 9 | 1299 | ||
NM_001282783.1 | NP_001269712.1 | 9 | 1240 | |||
NM_018006.4 | NP_060476.2 | 10 | 1406 | |||
NR_104240.1 | 10 | 1351 | ||||
NR_104241.1 | 9 | 1244 | ||||
XM_005261678.1 | XP_005261735.1 | 10 | 1034 | |||
XM_011530271.2 | XP_011528573.1 | 10 | 1169 | |||
XM_011530272.1 | XP_011528574.1 | 9 | 992 | |||
XM_011530274.2 | XP_011528576.1 | 9 | 1006 | |||
XM_011530275.1 | XP_011528577.1 | 10 | 990 | |||
GenBank mRNA | AB178028.1 | - | 10 | 1093 | ||
AK000212.1 | - | 1 | 1304 | |||
AK001002.1 | - | 10 | 1038 | |||
AK055295.1 | - | 2 | 1609 | |||
AK093743.1 | - | 2 | 2096 | |||
AK290712.1 | - | 10 | 1086 | |||
AY062123.2 | - | 10 | 1382 | |||
BC027991.1 | - | 9 | 898 | 5000797 | ||
DQ309998.1 | - | 9 | 1220 | |||
DQ309999.1 | - | 9 | 1275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
