siRNA Details
Gene Symbol: EFCAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014774.2 | NP_055589.1 | 11 | 3953 | ||
GenBank mRNA | AB007963.1 | - | 11 | 3953 | ||
AK001487.1 | - | 8 | 2452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
