siRNA Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040715.1 | NP_001035805.1 | 2, 3 | 1353 | ||
GenBank mRNA | AK094787.1 | - | 3, 4 | 690 | ||
AK096855.1 | - | 2, 3 | 145 | |||
AL834156.1 | - | 3, 4 | 1467 | |||
BC007594.1 | - | 1, 2 | 547 | |||
BC080183.1 | - | 2, 3 | 946 | 5742087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
